Manganese(III) oxide, 98%, Thermo Scientific Chemicals
Manganese(III) oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(III) oxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 087791.36
also known as 087791-36
Price (USD)/ Each
156.65
Online exclusive
174.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
156.65
Online exclusive
174.00 
Save 17.35 (10%)
Add to cart
Manganese(III) oxide, 98%, Thermo Scientific Chemicals
Catalog number087791.36
Price (USD)/ Each
156.65
Online exclusive
174.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1317-34-6
IUPAC Nameoxo[(oxomanganio)oxy]manganese
Molecular FormulaMn2O3
InChI KeyGEYXPJBPASPPLI-UHFFFAOYSA-N
SMILESO=[Mn]O[Mn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Manganese(III) oxide is used to produce ceramic magnets and semiconductors. Further, it serves as a precursor to lithium manganese oxide. In addition to this, it acts as a useful battery cathode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only