Thermo Scientific Chemicals

Tellurium(IV) iodide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 087795.06
5 g, Each
Thermo Scientific Chemicals

Tellurium(IV) iodide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 087795.06
5 g, Each
Quantity
Catalog number: 087795.06
also known as 087795-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-48-9
IUPAC Name
tetraiodo-λ⁴-tellane
Molecular Formula
I4Te
InChI Key
XCOKHDCPVWVFKS-UHFFFAOYSA-N
SMILES
I[Te](I)(I)I
Total Metal Impurities
1% max.

Description

Tellurium(IV) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text