Thermo Scientific Chemicals

Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals

Catalog number: 087839.06
5 g, Each
Thermo Scientific Chemicals

Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals

Catalog number: 087839.06
5 g, Each
Quantity
Catalog number: 087839.06
also known as 087839-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25895-60-7
IUPAC Name
sodium cyanoboranuide
Molecular Formula
CH3BNNa
InChI Key
CVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES
[Na+].[BH3-]C#N
Appearance (Color)
White to pale cream or pale yellow
Assay from Supplier's CofA
≥94.0 to ≤106.0% (Titration)
Form
Powder

Description

Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive; Highly flammable
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text