Thermo Scientific Chemicals

Deuterosulfuric acid, 96% min in D{2}O, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: 087857.18
50 g, Each
Thermo Scientific Chemicals

Deuterosulfuric acid, 96% min in D{2}O, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: 087857.18
50 g, Each
Quantity
Catalog number: 087857.18
also known as 087857-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13813-19-9
IUPAC Name
(²H₂)sulfuric acid
Molecular Formula
H2O4S
InChI Key
QAOWNCQODCNURD-ZSJDYOACSA-N
SMILES
[2H]OS(=O)(=O)O[2H]
Proton NMR
99.5% min
Concentration (weight/weight)
96% min

Description

Deuterated sulfuric acid when treated with benzene, the deuterium is used in order to observe the reaction course by NMR spectroscopy, deuterated sulfuric acid results in an electrophilic aromatic substitution of hydrogen by deuterium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text