Thermo Scientific Chemicals

Decaborane, 98%, Thermo Scientific Chemicals

Catalog number: 087892.03
1 g, Each
Thermo Scientific Chemicals

Decaborane, 98%, Thermo Scientific Chemicals

Catalog number: 087892.03
1 g, Each
Quantity
Catalog number: 087892.03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17702-41-9
Molecular Formula
B10H14
Molecular Weight (g/mol)
122.22
MDL Number
MFCD00011314
Form
crystalline powder and chunks
Appearance (Color)
White to pale yellow
Assay (unspecified)
> 97.5%

Description

Decaborane is used as a fuel for aneutronic fusion. It is also used in the preparation of vinyl decaborane as well as in the preparation of semiconductors that have low energy ion implantation. It is widely used in rocket propellants and serves as a catalyst in olefin polymerization reactions. It is a reactant for the preparation of carborane-conjugated quinoline carboxamide ligands of translocator protein for boron neutron capture therapy (BNCT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text