Zinc chloride, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals
Zinc chloride, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc chloride, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
25 g
100 g
Catalog number 087900.88
also known as 087900-88
Price (USD)/ Each
33.00
-
Add to cart
Quantity:
2.5 g
Request bulk or custom format
Price (USD)/ Each
33.00
Add to cart
Zinc chloride, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number087900.88
Price (USD)/ Each
33.00
-
Add to cart
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Zinc(II) chloride is used as a catalyst, as a dehydrating and condensing agent, soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in an organic synthesis, for example in the Friedel-crafts acylation, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only