Thermo Scientific Chemicals

Diphenylzinc, 98+%, Thermo Scientific Chemicals

Catalog number: 087908.03
1 g, Each
Thermo Scientific Chemicals

Diphenylzinc, 98+%, Thermo Scientific Chemicals

Catalog number: 087908.03
1 g, Each
Quantity
Catalog number: 087908.03
also known as 087908-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1078-58-6
Assay by NMR
> 98.0%
Appearance (Color)
White
Form
Crystals

Description

Diphenylzinc is used as the synthetic equivalent of ph- synthon. It is also useful in a Michael 1,4-addition reactions. It is used as a catalyst for propylene oxide polymerization in benzene medium. Further, it is a reactive coupling compound in Negishi cross-coupling reactions. In addition to this, it utilized in the preparation of arylcopper, arylsilver and arylgold compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text