Diphenylzinc, 98+%, Thermo Scientific Chemicals
Diphenylzinc, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylzinc, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 087908.06
also known as 087908-06
Price (USD)/ Each
536.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
536.00
Add to cart
Diphenylzinc, 98+%, Thermo Scientific Chemicals
Catalog number087908.06
Price (USD)/ Each
536.00
-
Add to cart
Chemical Identifiers
CAS1078-58-6
SpecificationsSpecification SheetSpecification Sheet
Assay by NMR> 98.0%
Appearance (Color)White
FormCrystals
Diphenylzinc is used as the synthetic equivalent of ph- synthon. It is also useful in a Michael 1,4-addition reactions. It is used as a catalyst for propylene oxide polymerization in benzene medium. Further, it is a reactive coupling compound in Negishi cross-coupling reactions. In addition to this, it utilized in the preparation of arylcopper, arylsilver and arylgold compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only