Thermo Scientific Chemicals

Thallium(III) trifluoroacetate, 95%, Thermo Scientific Chemicals

Catalog number: 087913.18
50 g, Each
Thermo Scientific Chemicals

Thallium(III) trifluoroacetate, 95%, Thermo Scientific Chemicals

Catalog number: 087913.18
50 g, Each
Quantity
Catalog number: 087913.18
also known as 087913-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23586-53-0
IUPAC Name
thallium(3+) tritrifluoroacetate
Molecular Formula
C6F9O6Tl
InChI Key
PSHNNUKOUQCMSG-UHFFFAOYSA-K
SMILES
[Tl+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Form
Solid
Assay from Suppliers CofA
≥94.0%

Description

Thallium(III) trifluoroacetate is an oxidizing agent used in the synthesis of S-substituted cysteine peptides and proteins containing disulfide bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text