Thermo Scientific Chemicals

Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 087920.AE
100 mL, Each
Thermo Scientific Chemicals

Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 087920.AE
100 mL, Each
Quantity
Catalog number: 087920.AE
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
Assay (Titration)
65-70%
Total Metal Impurities
0.001% max.

Description

In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuelsMain commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text