Thermo Scientific Chemicals

Molecular sieves, 13X, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog number: 087954.A1
1 kg, Each
Thermo Scientific Chemicals

Molecular sieves, 13X, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog number: 087954.A1
1 kg, Each
Quantity
Catalog number: 087954.A1
also known as 087954-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63231-69-6
IUPAC Name
Molecular sieves 13X
MDL Number
MFCD00131613
Appearance (Color)
White to pale yellow
Comment
Apparent density:≥37 lbs/ft³
Comment
CO₂ capacity at 250 Torr: ≥17 lbs/100 lbs
Loss on ignition
≤1.0% at 575°C
Form
Pellets

Description

Molecular sieves is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is preferred for static dehydration in closed liquid or gas systems, e.g., in packaging of drugs, electric components and perishable chemicals; water scavenging in printing and plastics systems and drying saturated hydrocarbon streams. Generally considered a universal drying agent in polar and nonpolar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text