Thermo Scientific Chemicals

Molecular sieves, 5A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog number: 087955.A1
1 kg, Each
Thermo Scientific Chemicals

Molecular sieves, 5A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog number: 087955.A1
1 kg, Each
Quantity
Catalog number: 087955.A1
also known as 087955-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69912-79-4
IUPAC Name
Molecular sieves 5A
MDL Number
MFCD00147627
Assay (unspecified)
Crush strength > 5 lbs

Description

Molecular sieves, 5A, 1-2mm (0.04-0.08in) dia. Pellets are used in desiccation and purification of air. It finds application in natural gas and liquid petroleum gas especially for dehydration and desulphurization. In chemistry laboratory, it is used for separating compounds and drying reaction starting materials. In addition, it is used as commercial adsorbents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text