Thermo Scientific Chemicals

Molecular sieves, 3A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog number: 087957.A1
1 kg, Each
Thermo Scientific Chemicals

Molecular sieves, 3A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog number: 087957.A1
1 kg, Each
Quantity
Catalog number: 087957.A1
also known as 087957-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
308080-99-1
IUPAC Name
Molecular sieves 3A
MDL Number
MFCD00131613
Appearance (Color)
Off-white
Assay (unspecified)
Bulk density: >/= 40 lb/cu ft
Assay (unspecified)
Equilibrium water capacity: > 19 wt % at 25 degC/ 17.5 mm Hg
Form
pellets

Description

Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for the drying of unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text