Chlorosulfonic acid, typically 99%, Thermo Scientific Chemicals
Chlorosulfonic acid, typically 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorosulfonic acid, typically 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
3 x 1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 087977.A1
also known as 087977-A1
Price (USD)/ Each
272.65
Online exclusive
303.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
272.65
Online exclusive
303.00 
Save 30.35 (10%)
Add to cart
Chlorosulfonic acid, typically 99%, Thermo Scientific Chemicals
Catalog number087977.A1
Price (USD)/ Each
272.65
Online exclusive
303.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7790-94-5
IUPAC Namesulfurochloridic acid
Molecular FormulaClHO3S
InChI KeyXTHPWXDJESJLNJ-UHFFFAOYSA-N
SMILESOS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Chlorosulfonic acid is used as detergent and as an anti-contrail agent. It is used as an intermediate in the production of other substances such as pharmaceuticals and chemicals. It finds application in producing smoke screens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only