This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 088056.AE
also known as 088056-AE
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.00
Save 5.35 (13%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.00
Save 5.35 (13%)
Add to cart
Cadmium, AAS standard solution, Specpure™ Cd 1000μg/mL, Thermo Scientific Chemicals
Chemical Name or MaterialCadmium, AAS standard solution
ConcentrationCd in 5% HNO3
Name NoteCd 1000μg/mL
Health Hazard 1H290-H315-H318-H350
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.