Chemical Name or MaterialCadmium, 1000μg/mL
Name NoteCd 1000μg/mL
Health Hazard 1Danger
Quantity500 mL
Health Hazard 2GHS H Statement
H350-H361-H314
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
View more