Thermo Scientific Chemicals
Cesium, AAS standard solution, Specpure™ Cs 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088058.AE
100 mL, Each
Thermo Scientific Chemicals
Cesium, AAS standard solution, Specpure™ Cs 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088058.AE
100 mL, Each
Quantity
Catalog number: 088058.AE
also known as 088058-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Cs2CO3