Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 088059.AE
also known as 088059-AE
Price (USD)/ Each
39.30
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
39.30
Add to cart
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Catalog number088059.AE
Price (USD)/ Each
39.30
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialChromium
Name NoteAAS Standard Solution
CAS7789-02-8
Health Hazard 1H314-H332-H335
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only