Copper, AAS standard solution, Specpure™ Cu 1000μg/mL, Thermo Scientific Chemicals
Copper, AAS standard solution, Specpure™ Cu 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper, AAS standard solution, Specpure™ Cu 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 088061.AP
also known as 088061-AP
Price (USD)/ Each
75.65
Online exclusive
84.30 
Save 8.65 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
84.30 
Save 8.65 (10%)
Add to cart
Copper, AAS standard solution, Specpure™ Cu 1000μg/mL, Thermo Scientific Chemicals
Catalog number088061.AP
Price (USD)/ Each
75.65
Online exclusive
84.30 
Save 8.65 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCopper
CAS7440-50-8
Health Hazard 1H290-H315-H318
Quantity500 mL
CAS7732-18-5
View more
Copper, AAS standard solution is used to determine copper in samples of water and water-suspendedsediment with a specific conductance not greater than 10,000 μS/cm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only