Thermo Scientific Chemicals
Gadolinium, AAS standard solution, Specpure™ Gd 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088065.AE
100 mL, Each
Thermo Scientific Chemicals
Gadolinium, AAS standard solution, Specpure™ Gd 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088065.AE
100 mL, Each
Quantity
Catalog number: 088065.AE
also known as 088065-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Gd2O3