Thermo Scientific Chemicals
Gold, AAS standard solution, Specpure™ Au 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088068.AP
500 mL, Each
Thermo Scientific Chemicals
Gold, AAS standard solution, Specpure™ Au 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088068.AP
500 mL, Each
Quantity
Catalog number: 088068.AP
also known as 088068-AP
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Au
Description
Gold, AAS standard solution is used for the determination of gold by flame atomic absorption spectrometry (FAAS) and graphite furnace atomic absorption spectrometry (GFAAS).