Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 088078.AP
also known as 088078-AP
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
Add to cart
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Catalog number088078.AP
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialManganese
CAS7439-96-5
Health Hazard 1H290-H315-H318
Quantity500 mL
CAS7697-37-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only