Thermo Scientific Chemicals
Potassium, AAS standard solution, Specpure™ K 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088087.AP
500 mL, Each
Thermo Scientific Chemicals
Potassium, AAS standard solution, Specpure™ K 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 088087.AP
500 mL, Each
Quantity
Catalog number: 088087.AP
also known as 088087-AP
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000 µg/ml Potassium
Concentration
1,000?g/mL ?1%
Starting Material
KNO3