Thermo Scientific Chemicals

Iron boride, 98%, Thermo Scientific Chemicals

Catalog number: 088146.14
25 g, Each
Thermo Scientific Chemicals

Iron boride, 98%, Thermo Scientific Chemicals

Catalog number: 088146.14
25 g, Each
Quantity
Catalog number: 088146.14
also known as 088146-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12006-84-7
Assay (unspecified)
97.6% min

Description

Iron boride is used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. Being inexpensive as well as having excellent mechanical, frictional and corrosion resistant properties, it can be used as replacement of ceramic or cermet type of materials in fabrication and machining. It is also involved in the production of borated steels and borated stainless steels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text