Thermo Scientific Chemicals

Magnesium boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 088149.22
100 g, Each
Thermo Scientific Chemicals

Magnesium boride, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 088149.22
100 g, Each
Quantity
Catalog number: 088149.22
also known as 088149-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12007-25-9
IUPAC Name
diboron magnesium
Molecular Formula
B2Mg
InChI Key
PZKRHHZKOQZHIO-UHFFFAOYSA-N
SMILES
[B].[B].[Mg]
Total Metal Impurities
1% max.

Description

Magnesium Boride exhibits super conducting properties and utilized in MRI superconducting magnet, which is a closed-loop cryocooler. It is used as propellants, pyrotechnics and explosives. It can be used as high temperature boron-based thermoelectric materials. It is also used in superconducting transformers, rotors, transmission cables, superconducting radio frequency cavities to minimize energy loss.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text