Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals
Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 088220.14
also known as 088220-14
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
Add to cart
Strontium oxide, 99.5% (metals basis), SrO ^=97%, Thermo Scientific Chemicals
Catalog number088220.14
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-11-0
IUPAC Namestrontium(2+) oxidandiide
Molecular FormulaOSr
InChI KeyIATRAKWUXMZMIY-UHFFFAOYSA-N
SMILES[O--].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only