Zinc selenide, Optical Grade, 99.999% (metals basis), Thermo Scientific Chemicals
Zinc selenide, Optical Grade, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc selenide, Optical Grade, 99.999% (metals basis), Thermo Scientific Chemicals

For vacuum deposition
Have Questions?
Quantity:
25 g
Catalog number 088298.14
also known as 088298-14
Price (USD)/ Each
91.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
91.80
Add to cart
Zinc selenide, Optical Grade, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number088298.14
Price (USD)/ Each
91.80
-
Add to cart
Chemical Identifiers
CAS1315-09-9
IUPAC Namezinc(2+) selandiide
Molecular FormulaSeZn
InChI KeyGTLQJUQHDTWYJC-UHFFFAOYSA-N
SMILES[Zn++].[Se--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only