Thermo Scientific Chemicals

Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals

Catalog number: 088300.JQ
0.1 mol, Each
Thermo Scientific Chemicals

Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals

Catalog number: 088300.JQ
0.1 mol, Each
Quantity
Catalog number: 088300.JQ
also known as 088300-JQ
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4984-82-1
IUPAC Name
sodium cyclopenta-2,4-dien-1-ide
Molecular Formula
C5H5Na
InChI Key
OHUVHDUNQKJDKW-UHFFFAOYSA-N
SMILES
[Na+].[CH-]1C=CC=C1
Appearance (Color)
Pink to red
Concentration
2.0-3.0M
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text