Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals
Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals

Reagent for the preparation of metallocenes
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mol
0.1 mol
Catalog number 088300.JT
also known as 088300-JT
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
0.5 mol
Request bulk or custom format
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
Add to cart
Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals
Catalog number088300.JT
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4984-82-1
IUPAC Namesodium cyclopenta-2,4-dien-1-ide
Molecular FormulaC5H5Na
InChI KeyOHUVHDUNQKJDKW-UHFFFAOYSA-N
SMILES[Na+].[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to red
Concentration2.0-3.0M
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only