Aluminum Silicon powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
Aluminum Silicon powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum Silicon powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 088322.36
also known as 088322-36
Price (USD)/ Each
218.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
218.00
Add to cart
Aluminum Silicon powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
Catalog number088322.36
Price (USD)/ Each
218.00
-
Add to cart
Chemical Identifiers
CAS11145-27-0
IUPAC Namealuminium silicon
Molecular FormulaAlSi
InChI KeyCSDREXVUYHZDNP-UHFFFAOYSA-N
SMILES[Al].[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Elemental AnalysisSi: 11.5-12.5%
Appearance (Color)Grey
Total Metal Impurities≤1.0%
Aluminum Silicon powder is a reducing agent which is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only