Thermo Scientific Chemicals
Titanium Nickel powder composite, -325 mesh, Thermo Scientific Chemicals
Catalog number: 088384.22
100 g, Each
Thermo Scientific Chemicals
Titanium Nickel powder composite, -325 mesh, Thermo Scientific Chemicals
Catalog number: 088384.22
100 g, Each
Quantity
Catalog number: 088384.22
also known as 088384-22
Price (USD)
Have Questions?
Particle Size
-325 mesh (less than 45 micron)
Description
A 70% titanium, 30% nickel composite powder strained through -325 mesh. Also used in alloy powder, bone staples and antioxidant coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov