Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 088390.22
also known as 088390-22
Price (USD)/ Each
61.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
61.70
Add to cart
Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
Catalog number088390.22
Price (USD)/ Each
61.70
-
Add to cart
Specifications
Chemical Name or MaterialStainless Steel powder, Type 316-L
ColorGray
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only