Thermo Scientific Chemicals
Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
Catalog number: 088390.A3
2 kg, Each
Thermo Scientific Chemicals
Stainless Steel powder, -325 mesh, Type 316-L, Thermo Scientific Chemicals
Catalog number: 088390.A3
2 kg, Each
Quantity
Catalog number: 088390.A3
also known as 088390-A3
Price (USD)
Quantity
-
Have Questions?
Description
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov