Thermo Scientific Chemicals

Titanium aluminide, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 088393.09
10 g, Each
Thermo Scientific Chemicals

Titanium aluminide, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 088393.09
10 g, Each
Quantity
Catalog number: 088393.09
also known as 088393-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12003-96-2
Form
Powder
Particle Size
ca. 325 Mesh
Appearance (Color)
Silver grey
Assay from Supplier's CofA
≥99.5%

Description

It finds use in several applications including automobiles and aircraft. Also used in air frames, investment casting, turbocharger rotors, and wire/fiber metal matrix composites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text