Thermo Scientific Chemicals

Gallium arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 088458.04
2 g, Each
Thermo Scientific Chemicals

Gallium arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 088458.04
2 g, Each
Quantity
Catalog number: 088458.04
also known as 088458-04
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1303-00-0
IUPAC Name
gallanylidynearsane
Molecular Formula
AsGa
InChI Key
JBRZTFJDHDCESZ-UHFFFAOYSA-N
SMILES
[Ga]#[As]
Total Metal Impurities
0.001% max.

Description

Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text