Thermo Scientific Chemicals

Indium antimonide, Electronic Grade, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 088460.18
50 g, Each
Thermo Scientific Chemicals

Indium antimonide, Electronic Grade, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 088460.18
50 g, Each
Quantity
Catalog number: 088460.18
also known as 088460-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-41-0
Total Metal Impurities
0.01% max.

Description

Indium antimonide is used in semiconductor electronics, as an infrared detector and filter. Used in thermal imaging cameras, FLIR systems, infrared homing missile guidance systems, photo-Dember emitter and in infrared astronomy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text