Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.
Column Format | Guard Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 10 mm |
Max. Pressure | 6000 psi (415 bar) |
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 100 mm |
Max. Pressure | 6000 psi (415 bar) |
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 250 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Preparative Column, Standard |
Diameter (Metric) | 7.8 mm |
Length (Metric) | 100 mm |
Max. Pressure | 5800 psi (400 bar) |
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Alkyl Amide |
Product Line | MAbPac |
Packing Material | Spherical, High Purity Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
088482 also known as 088482 | Each | 5 μm | 4.6 mm | 10 mm | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088480 | Each | 5 μm | 4.6 mm | 100 mm | 1,513.90 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088481 | Each | 5 μm | 4.6 mm | 250 mm | 2,195.05 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP6997 | Each | 5 μm | 7.8 mm | 100 mm | Request A Quote | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Unique Chemistry Designed for Optimal Selectivity of mAbs
The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.