Thermo Scientific Chemicals

Aluminum chloride, anhydrous, 99.985% (metals basis), Thermo Scientific Chemicals

Catalog number: 088488.30
250 g, Each
Thermo Scientific Chemicals

Aluminum chloride, anhydrous, 99.985% (metals basis), Thermo Scientific Chemicals

Catalog number: 088488.30
250 g, Each
Quantity
Catalog number: 088488.30
also known as 088488-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-70-0
IUPAC Name
aluminium(3+) trichloride
Molecular Formula
AlCl3
InChI Key
VSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES
[Al+3].[Cl-].[Cl-].[Cl-]
Form
Powder
Appearance (Color)
White
Residual water
< 2000 ppm
Total Metal Impurities
≤0.015% (150 ppm)

Description

Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text