Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals
Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 088517.09
also known as 088517-09
Price (USD)/ Each
206.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
243.00 
Save 36.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
206.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
243.00 
Save 36.35 (15%)
Add to cart
Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals
Catalog number088517.09
Price (USD)/ Each
206.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
243.00 
Save 36.35 (15%)
-
Add to cart
Chemical Identifiers
CAS12005-86-6
IUPAC Nameλ⁵-arsenic(5+) sodium hexafluoride
Molecular FormulaAsF6Na
InChI KeyIYPUMRHORYWLBJ-UHFFFAOYSA-H
SMILES[F-].[F-].[F-].[F-].[F-].[F-].[Na+].[As+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Sodium hexafluoroarsenate(V) is generally used for pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only