Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use the Thermo Scientific™ MAbPac™ HIC-20 HPLC column for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity. The advanced surface bonding technology provides excellent chemical stability, high resolution, and unique selectivity optimized for mAbs and different from other HIC phases.
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 100 mm |
Max. Pressure | 6000 psi (415 bar) |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Spherical, High Purity Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 250 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Spherical, High Purity Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Guard Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 10 mm |
Max. Pressure | 6000 psi (415 bar) |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Spherical, High Purity Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Preparative Column, Standard |
Diameter (Metric) | 10 mm |
Length (Metric) | 150 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Alkyl Amide |
Product Line | MAbPac |
Packing Material | Spherical, High Purity Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Unit Size | Each |
Column Format | Preparative Column, Standard |
Diameter (Metric) | 10 mm |
Length (Metric) | 250 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 1000 Å |
Temperature | 60°C |
Stationary Phase | Alkyl Amide |
Product Line | MAbPac |
Packing Material | Spherical, High Purity Silica |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
088553 | Each | 5 μm | 4.6 mm | 100 mm | 1,513.90 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088554 | Each | 5 μm | 4.6 mm | 250 mm | 2,195.05 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088555 | Each | 5 μm | 4.6 mm | 10 mm | 250.10 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP6961 | Each | 5 μm | 10 mm | 150 mm | Request A Quote | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP6995 | Each | 5 μm | 10 mm | 250 mm | Request A Quote | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
This column is based on wide-pore silica particles with proprietary alkyl amide groups offering: