Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use the Thermo Scientific™ MAbPac™ HIC-Butyl HPLC columns to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups. The column's hydrophilic resin provides excellent biocompatibility and low carryover, while the optimal density of the butyl groups provides high resolution separation.
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 100 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Hydrophilic Polymer |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Guard Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 10 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | 60°C |
Stationary Phase | Silica |
Product Line | MAbPac |
Packing Material | Hydrophilic Polymer |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Preparative Column, Standard |
Diameter (Metric) | 7.8 mm |
Length (Metric) | 100 mm |
Max. Pressure | 4000 psi (275 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | 60°C |
Stationary Phase | Polyamide |
Product Line | MAbPac |
Packing Material | Hydrophilic Polymer |
Column Type | Hydrophobic Interaction |
Phase | Hydrophobic Interaction |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
088558 | Each | 5 μm | 4.6 mm | 100 mm | 1,513.90 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088559 also known as 088559 | Each | 5 μm | 4.6 mm | 10 mm | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP6965 | Each | 5 μm | 7.8 mm | 100 mm | Request A Quote | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|