Thermo Scientific Chemicals

Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, Thermo Scientific Chemicals

Catalog number: 088568.LQ
1 Unit, Each
Thermo Scientific Chemicals

Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, Thermo Scientific Chemicals

Catalog number: 088568.LQ
1 Unit, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 088568.LQ
also known as 088568-LQ
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-93-9
IUPAC Name
sulfuric acid
Molecular Formula
H2O4S
InChI Key
QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
1.006-2.004N
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text