Thermo Scientific Chemicals

Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals

Catalog number: 088614.LS
6 Units, Each
Thermo Scientific Chemicals

Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals

Catalog number: 088614.LS
6 Units, Each
Quantity
Catalog number: 088614.LS
also known as 088614-LS
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7761-88-8
IUPAC Name
silver(1+) nitrate
Molecular Formula
AgNO3
InChI Key
SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES
[Ag+].[O-][N+]([O-])=O
Concentration
Final Concentration after dilution to 1 Liter, 0.1N

Description

Silver nitrate 0.1N standard volumetric solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text