Thermo Scientific Chemicals

Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L, Thermo Scientific Chemicals

Catalog number: 088632.LQ
1 U, Each
Thermo Scientific Chemicals

Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L, Thermo Scientific Chemicals

Catalog number: 088632.LQ
1 U, Each
Quantity
Catalog number: 088632.LQ
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-84-9, 7732-18-5
Form
Liquid
Comment
Final concentration 1%

Description

Starch indicator 1% solution is used as an indicator for redox titrations. It is also used in the detection of end point in iodometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text