Search Thermo Fisher Scientific
Compatible with a wide variety of applications, Dionex DRS 600 suppressors have simple on/off control, improved gradient performance, and are enabled for IC-MS.
Catalog Number | Description |
---|---|
088667 | Dionex ADRS 600 Anion Dynamically Regenerated Suppressor (2 mm) (Replacement for P/N 082541 Dionex AERS Anion Electrolytically Regenerated Suppressor (2 mm)) |
088666 | Dionex ADRS 600 Anion Dynamically Regenerated Suppressor (4 mm) (Replacement for P/N 082540 Dionex AERS Anion Electrolytically Regenerated Suppressor (4 mm)) |
088668 | Dionex CDRS 600 Cation Dynamically Regenerated Suppressor (4 mm) (Replacement for P/N 082542 Dionex CERS Cation Electrolytically Regenerated Suppressor (4 mm)) |
088670 | Dionex CDRS 600 Cation Dynamically Regenerated Suppressor (2 mm) (Replacement for P/N 082543 Dionex CERS Cation Electrolytically Regenerated Suppressor (2 mm)) |
Please note: The Dionex DRS 600 suppressors will be discontinued on December 31st, 2025. The Thermo Scientific™ Dionex™ NGES Next Generation Electrolytic Suppressor is the recommended replacement. For more information, please visit the Dionex NGES Electrolytic Suppressor page.
Use the Thermo Scientific™ Dionex™ DRS 600 Dynamically Regenerated Suppressor for electrolytically regenerated suppression in ion chromatography (IC) applications requiring high capacity, low noise, high backpressure resiliency, and fast startup. This suppressor is a fundamental component of a reagent-free ion chromatography (RFIC™) system.
Key features
The Dionex DRS 600 Dynamically Regenerated Suppressor is the recommended replacement for the Thermo Scientific™ Dionex™ ERS™ 500 Electrolytically Regenerated Suppressor.
Share catalog number, name or link