Search Thermo Fisher Scientific
Compatible with a wide variety of applications, Dionex DRS 600 suppressors have simple on/off control, improved gradient performance, and are enabled for IC-MS.
Catalog Number | Description |
---|---|
088668 | Dionex CDRS 600 Cation Dynamically Regenerated Suppressor (4 mm) (Replacement for P/N 082542 Dionex CERS Cation Electrolytically Regenerated Suppressor (4 mm)) |
088666 | Dionex ADRS 600 Anion Dynamically Regenerated Suppressor (4 mm) (Replacement for P/N 082540 Dionex AERS Anion Electrolytically Regenerated Suppressor (4 mm)) |
088667 | Dionex ADRS 600 Anion Dynamically Regenerated Suppressor (2 mm) (Replacement for P/N 082541 Dionex AERS Anion Electrolytically Regenerated Suppressor (2 mm)) |
088670 | Dionex CDRS 600 Cation Dynamically Regenerated Suppressor (2 mm) (Replacement for P/N 082543 Dionex CERS Cation Electrolytically Regenerated Suppressor (2 mm)) |
Use the Thermo Scientific™ Dionex™ DRS 600 Dynamically Regenerated Suppressor for electrolytically regenerated suppression in ion chromatography (IC) applications requiring high capacity, low noise, high backpressure resiliency, and fast startup. This suppressor is a fundamental component of a reagent-free ion chromatography (RFIC™) system.
The Dionex ADRS 600 Anion Dynamically Regenerated Suppressor allows external water mode operation and enables analyses requiring IC coupled with mass spectrometry (MS).
The Dionex DRS 600 Dynamically Regenerated Suppressor is a direct replacement for the Thermo Scientific™ Dionex™ ERS™ 500 Electrolytically Regenerated Suppressor.