Silver trifluoromethanesulfonate, 98%
Silver trifluoromethanesulfonate, 98%
Silver trifluoromethanesulfonate, 98%
Silver trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Silver trifluoromethanesulfonate, 98%

CAS: 2923-28-6 | CAgF3O3S | 256.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 088722.04
also known as 088722-04
Price (USD)
30.10
Each
In stock
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)
30.10
Each
Add to cart
Chemical Identifiers
CAS2923-28-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Ask our AI about this Product
Silver trifluoromethanesulfonate is useful in halide abstraction. It helps in activating sulphur protecting groups at the anomeric centres of carbohydrates in the presence of iodine. It is employed as a triflating agent in combination with Palladium compounds, as a catalyst for cross coupling reactions, and in several organic transformation reactions. Polyurethanes prepared from silver triflate exhibit high permeability and selectivity towards olefins in their separation from paraffin mixtures. Polyethylene oxide - silver triflate composites find application in humidity sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0463793Certificate of AnalysisAug 30, 2024088722.09, 088722.18, 088722.04
I12M24Certificate of AnalysisDec 08, 2021088722.09, 088722.18, 088722.04
10190668Certificate of AnalysisDec 08, 2021088722.09, 088722.18, 088722.04
10190039Certificate of AnalysisDec 08, 2021088722.09, 088722.18, 088722.04
F27R006Certificate of AnalysisDec 08, 2021088722.09, 088722.18, 088722.04
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection