Thermo Scientific Chemicals

Silver trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 088722.09
10 g, Each
Thermo Scientific Chemicals

Silver trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 088722.09
10 g, Each
Quantity
Catalog number: 088722.09
also known as 088722-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2923-28-6
Assay (unspecified)
97.6% min

Description

Silver trifluoromethanesulfonate is useful in halide abstraction. It helps in activating sulphur protecting groups at the anomeric centres of carbohydrates in the presence of iodine. It is employed as a triflating agent in combination with Palladium compounds, as a catalyst for cross coupling reactions, and in several organic transformation reactions. Polyurethanes prepared from silver triflate exhibit high permeability and selectivity towards olefins in their separation from paraffin mixtures. Polyethylene oxide - silver triflate composites find application in humidity sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text