Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 150 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 3 μm |
pH | 0.5 to 10 |
Pore Size | 120 Å |
Temperature | <80°C |
USP Type | L1 |
Stationary Phase | C18 |
Product Line | Acclaim |
Packing Material | Spherical, High Purity Silica |
Column Type | Aminoglycoside Analysis |
Phase | Reversed Phase |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 150 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 3 μm |
pH | 0.5 to 10 |
Pore Size | 120 Å |
Temperature | <80°C |
USP Type | L1 |
Stationary Phase | C18 |
Product Line | Acclaim |
Packing Material | Spherical, High Purity Silica |
Column Type | Aminoglycoside Analysis |
Phase | Reversed Phase |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 3 mm |
Flow Rate | 0.425 mL/min |
Length (Metric) | 150 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 3 μm |
pH | 0.5 to 10 |
Pore Size | 120 Å |
Temperature | <80°C |
USP Type | L1 |
Stationary Phase | C18 |
Product Line | Acclaim |
Packing Material | Spherical, High Purity Silica |
Column Type | Aminoglycoside Analysis |
Phase | Reversed Phase |
Unit Size | Each |
Column Format | Guard Column |
Diameter (Metric) | 2.1 mm |
Length (Metric) | 10 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 3 μm |
pH | 0.5 to 10 |
Pore Size | 120 Å |
Quantity | 2 Guard Cartridges/Pk. (Requires Guard Cartridge Holder 069580) |
Temperature | <80°C |
USP Type | L1 |
Stationary Phase | C18 |
Product Line | Acclaim |
Packing Material | Spherical, High Purity Silica |
Column Type | Aminoglycoside Analysis |
Phase | Reversed Phase |
Unit Size | Pack of 2 |
Column Format | Guard Column |
Diameter (Metric) | 3 mm |
Length (Metric) | 10 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 3 μm |
pH | 0.5 to 10 |
Pore Size | 120 Å |
Quantity | 2 Guard Cartridges/Pk. (Requires Guard Cartridge Holder 069580) |
Temperature | <80°C |
USP Type | L1 |
Stationary Phase | C18 |
Product Line | Acclaim |
Packing Material | Spherical, High Purity Silica |
Column Type | Aminoglycoside Analysis |
Phase | Reversed Phase |
Unit Size | Pack of 2 |
Column Format | Guard Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 10 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 3 μm |
pH | 0.5 to 10 |
Pore Size | 120 Å |
Quantity | 2 Guard Cartridges/Pk. (Requires Guard Cartridge Holder 069580) |
Temperature | <80°C |
USP Type | L1 |
Stationary Phase | C18 |
Product Line | Acclaim |
Packing Material | Spherical, High Purity Silica |
Column Type | Aminoglycoside Analysis |
Phase | Reversed Phase |
Unit Size | Pack of 2 |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
088757 | Each | 3 μm | 4.6 mm | 150 mm | 1,051.12 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088753 | Each | 3 μm | 2.1 mm | 150 mm | Request A Quote | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088755 | Each | 3 μm | 3 mm | 150 mm | 1,051.12 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088754 | Pack of 2 | 3 μm | 2.1 mm | 10 mm | Request A Quote | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088756 | Pack of 2 | 3 μm | 3 mm | 10 mm | 333.87 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088758 | Pack of 2 | 3 μm | 4.6 mm | 10 mm | 274.43 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Acclaim AmG C18 column is specifically designed for ion-pairing reversed-phase HPLC (IP-RPLC) analysis of aminoglycoside antibiotics using volatile perfluorinated carboxylic acids as the ion-pairing reagent, such as 100 mM trifluoroacetic acid (TFA). The Acclaim AmG C18 stationary phase is based on a proprietary C18 bonding technology that provides extremely high stability towards acidic conditions. This specialty column exhibits excellent selectivity and high resolution for the aminoglycoside antibiotics analysis. In addition, it is easy to use because simple aqueous mobile phase (e.g. 100 mM TFA) without organic solvent and pH adjusting buffer is sufficient in most cases.
The column can be used with UV, CAD, ELSD detectors