Thermo Scientific Chemicals

Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 088944.36
500 g, Each
Thermo Scientific Chemicals

Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 088944.36
500 g, Each
Quantity
Catalog number: 088944.36
also known as 088944-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10039-54-0
IUPAC Name
bis(hydroxyazanium) sulfate
Molecular Formula
H8N2O6S
InChI Key
VGYYSIDKAKXZEE-UHFFFAOYSA-L
SMILES
[NH3+]O.[NH3+]O.[O-]S([O-])(=O)=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay (unspecified)
≥98.5%
Assay (unspecified)
Chloride: <0.0002%
Assay (unspecified)
Heavy Metal: <0.0002%

Description

Hydroxylammonium sulfate is used to produce hydroxylamine-O-sulfonic acid from chlorosulfuric acid. It is widely used in organic synthesis for the conversion of aldehydes and ketones to oximes, carboxylic acids and their derivatives (e.g. esters) to hydroxamic acids, isocyanates to N-hydroxyureas and nitriles to amidoximes. It acts as a radical scavenger which is used to terminate the radical polymerization reactions. It finds application as an antioxidant in natural rubber. It is also used in photography as a stabilizer for color developers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text